Nalgene™ Wide-Mouth HDPE Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth HDPE Packaging Bottles with Closure: Bulk Pack
Actual product may vary
Nalgene™ Wide-Mouth HDPE Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth HDPE Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth HDPE Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth HDPE Packaging Bottles with Closure: Bulk Pack
Confidently ship and store these tough plastic bottles, which are easy to fill, chemical-resistant, and leakproof when use with Nalgene closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
312189-00321000 mL63-415 mm
FKS1037424330 mL28-415 mm
FIS03-313-15B60 mL28-415 mm
312189-0004125 mL38-415 mm
FIS03-313-15C125 mL38-415 mm
FIS03-313-15D250 mL43-415 mm
312189-0016500 mL53-415 mm
FIS03-313-15E500 mL53-415 mm
FIS03-313-15F1000 mL63-415 mm
Catalog number 312189-0032
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Closure Size:
63-415 mm
Confidently ship and store these Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Packaging Bottles, which are easy to fill and resistant to chemicals. These wide-mouthed, high-density polyethelyene bottles are injection blow-molded for strength and uniform wall thickness.
Features:
  • Tough plastic construction
  • Made from high-purity, food-grade resins
  • Polypropylene closure
  • Leakproof† when used with Nalgene closures
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorTranslucent
DescriptionWide-Mouth Packaging Bottles
Diameter (Metric) Inner Neck52 mm
MaterialHDPE
No. per Case50
No. per Pack50
Closure ColorNatural, translucent
Neck Finish63 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)199 mm
Product TypeWide Mouth Packer Bottle
Width (English)3.6 in.
Height (English)7.83 in.
Closure MaterialPolypropylene
Closure Size63-415 mm
Diameter (Metric)91 mm
Closure TypeScrew (Bulk Packed, Unassembled)
Through the Closure DispensingNo
Unit SizeCS