Kimble™ Vacuum Desiccator with Collar, 250 x 330
Kimble™ Vacuum Desiccator with Collar, 250 x 330
Actual product may vary
Kimble™ Vacuum Desiccator with Collar, 250 x 330
Ideal for removing moisture from solids, storage of hygroscopic chemicals, and applications requiring vacuum desiccation.
Have Questions?
Catalog number 3120-0250
Price (SGD)
57.00
PK
Add to cart
Price (SGD)
57.00
PK
Add to cart
  • Greased rotating collar valve is used to facilitate vacuum desiccation
  • Includes solid glass lid with collar valve, metal plate, and a recessed base for desiccant containment
  • Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II requirements
  • Can be used for cooling oven-dried samples while maintaining sample dryness
Specifications
Product TypeVacuum Desiccator with Collar
Unit SizePK
Frequently asked questions (FAQs)
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.