Skullerz™ Odin Safety Glasses/Sunglasses
Skullerz™ Odin Safety Glasses/Sunglasses
Actual product may vary
Skullerz™ Odin Safety Glasses/Sunglasses
Full-frame, protective work safety sunglasses feature tough scratch-resistant lenses that filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun.
Have Questions?
Catalog NumberFrame ColorLens Tint
3115-0050BlackYellow
Catalog number 3115-0050
Price (SGD)
1,596.00
CS
Add to cart
Frame Color:
Black
Lens Tint:
Yellow
Price (SGD)
1,596.00
CS
Add to cart
  • Durable frames bend and flex without breaking
  • Non-slip rubber temples keep the glasses secure and resist sweat
  • Integrated accessory receptors at each temple improve airflow when not in use.
  • For added safety, an elastic retainer strap or foam gasket insert (both sold separately) can be attached to the temples.
  • Available with polarized lenses or with anti-fogging solution
  • Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments.
  • Anti-glare glasses block the harsh reflected light from water, keeping eyes comfortable around glare.
  • Fog-Off™ treatment won't wash off or wear off and is ideal for use with dust masks.
  • Certifications and Testing:
    • Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
    • Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
  • Ideal for protecting eyes during outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
  • Available in a variety of lens options ideally suited for a variety of tasks and light environments.
Specifications
Lens CoatingImpact-resistant, Scratch-resistant, UV-treated
Certifications/ComplianceANSI Z87.1+, CSA Z94.3 and MIL-PRF 32432 Ballistic Impact
Lens TintYellow
For Use With (Application)Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical
Frame ColorBlack
No. per Case12/Cs.
Unit SizeCS