Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes
Actual product may vary
Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes
Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes
Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes
Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes
Improve rotor fit and performance using these high-speed round-bottom PPCO centrifuge tubes, featuring uniform wall thickness and diameter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)
3110-038038 mL88.1 mm25.2 mm
3110-015015 mL112.8 mm15.8 mm
FIS05-562-10D15 mL112.8 mm15.8 mm
3110-050050 mL102.4 mm28.5 mm
FKS1046003350 mL102.4 mm28.5 mm
3110-1000100 mL163.1 mm31.5 mm
Catalog number 3110-0380
HSN 39269099
Price (SGD)
38.00
Special offer
Ends: 30-Jun-2026
54.00
Save 16.00 (30%)
PK
Capacity (Metric):
38 mL
Length (Metric):
88.1 mm
Diameter (Metric):
25.2 mm
Price (SGD)
38.00
Special offer
Ends: 30-Jun-2026
54.00
Save 16.00 (30%)
PK
Improve rotor fit and performance using Thermo Scientific™ Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes, featuring uniform wall thickness and diameter. Tubes have excellent chemical resistance.
Features
  • Round bottoms that fit high-speed rotors in refrigerated or nonrefrigerated centrifuges
  • Tubes must be filled at least 80% of total capacity for proper performance
  • For use to 50,000 x g in all refrigerated and non-refrigerated centrifuges (used with DS3111-series friction fit caps for proper performance)
  • Molding technique for uniform wall thickness and increased strength
  • Lip on Part Nos. 3110-1000 and 3110-9500 allows easier retrieval from rotor cavities
  • Uniform diameter (no taper) for better rotor fit and force tolerance
  • Economical choice—may also be used as test tubes
  • Longer lasting than polycarbonate tubes under conditions of repeated autoclaving
  • Autoclavable 
Ordering Information:
Supplied without closure; friction-fit closures available separately (DS3111-series)
12ml tube uses the 16mm closure
15ml tube uses the 16mm closure
16ml tube uses the 18mm closure
38ml tube uses the 25mm closure
50ml tubes use the 29mm closure
100ml tubes use the 32mm closure
Recycling Information: 
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.
.
Specifications
Capacity (Metric)38 mL
MaterialPolypropylene Copolymer
No. per Case100
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)88.1 mm
Bottom ShapeRound
Writing AreaNo
Diameter (Metric)25.2 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizePK
Frequently asked questions (FAQs)
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.