Test Tube Racks for MaxQ™ Shaker
Test Tube Racks for MaxQ™ Shaker
Actual product may vary
Test Tube Racks for MaxQ™ Shaker
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHoldsArray
30183OrangeTubes (14 to 16 mm), Centrifuge Tubes (15 mL)6 x 6
30180BIRedTubes (10 to 13 mm)6 x 12
30182
also known as GT30182
OrangeTubes (14 to 16 mm), Centrifuge Tubes (15 mL)6 x 12
30184
also known as GT30184
WhiteTubes (17 to 20 mm)4 x 10
30187
also known as A30187
BlueTubes (21 to 25 mm)4 x 4
30188
also known as GT30188
GreenTubes (26 to 30 mm)3 x 8 array
30189
also known as GT30189
GreenTubes (26 to 30 mm), Centrifuge Tubes (50 mL)3 x 3
Catalog number 30183
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Orange
Holds:
Tubes (14 to 16 mm), Centrifuge Tubes (15 mL)
Array:
6 x 6
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
Specifications
Product TypeTest Tube Rack
DescriptionOrange; 14 to 16 mm; 6 x 6 array (fits 15 mL centrifuge tubes)
Array6 x 6
ColorOrange
HoldsTubes (14 to 16 mm), Centrifuge Tubes (15 mL)
Positions36
For Use With (Equipment)MaxQ™ Shaker Platform
Unit SizeEA