Hughes Laboratory Drench Shower
Hughes Laboratory Drench Shower
Actual product may vary
Hughes Laboratory Drench Shower
Hughes Laboratory Drench Shower is ideal for labs, schools and general light duty indoor use where space is at a premium, this emergency drench shower is suitable for use in applications that do not require an eye wash and are not subject to the possibility of freezing or overheating.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMountingHeight (English)
30106
also known as R30106
Ring Main14 in.
30105
also known as GB30105
Ceiling14 in.
Catalog number 30106
also known as R30106
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Mounting:
Ring Main
Height (English):
14 in.
Features:
  • Specially designed valve with low operating torque offering reliable, easy on/off operation when emergencies occur.
  • Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure.
  • Low operating torque offers reliable, easy on/off operation.
  • Durable, coated stainless steel pull handle with nylon valve connection is easy to grip and activate for immediate water drenching from the shower rose that is designed for high flow rates.
  • Includes an inspection record tag for recording test and service history
  • Powder coated stainless steel pipework and rose ensures resistance to corrosive and flammable materials
  • Easy to clean
  • High-performance quarter-turn ball valve made of 316L stainless steel endure the harshest environments and site conditions
  • Includes a universal eye wash and safety shower sign for enhanced visibility
  • Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements
Specifications
ActivationPull Handle
Certifications/ComplianceANSI Z358.1-2014 and EN15154 Compliant
ColorGreen Powder Coat
Depth (English)14 in.
Depth (Metric)356 mm
For Use With (Application)Emergency Shower and Eye Wash with Water
Height (English)14 in.
Height (Metric)356 mm
Includes1 Unit
Inlet Connection (Metric)1 in. (F) NPT
MaterialPowder-coated Stainless Steel
Material (Piping Connection)Stainless Steel
MountingRing Main
Pressure22 to 87 psi
Product TypeSafety Shower and Emergency Eye Wash
Width (English)26 in.
Width (Metric)660 mm
Unit SizeEA