Nalgene™ Reusable Filter Units
Nalgene™ Reusable Filter Units
Actual product may vary
Nalgene™ Reusable Filter Units
Eliminate the need for a manifold or breakable glass vacuum flask. Nalgene Reusable Filter Holders with Receiver are complete, environmentally-friendly systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric) Receiver
300-41001000 mL
300-4000250 mL
FIS09-740-23A250 mL
300-4050500 mL
FIS09-740-23D500 mL
FIS09-740-23E1000 mL
FKS101692701000 mL
Catalog number 300-4100
Price (SGD)
330.00
EA
Add to cart
Volume (Metric) Receiver:
1000 mL
Price (SGD)
330.00
EA
Add to cart
Reusable filter units include two membrane support plates: a clear one-piece plate for sterilization applications and a two-piece white plate for analytical applications. Accepts 47 or 50 mm diameter membranes. Two side-arms allow connection to vacuum line. Tubing adapters accept 1/4- to 5/16-inch (6 to 8 mm) I.D. vacuum tubing. 4 units per case.
Features
  • Replaces metal or breakable glass – major components are made of durable, transparent, autoclavable polysulfone
  • Use for sterilization or analysis. Filter holders come with sterilization and analytical membrane support plates
  • Designed to be used with vacuum or pressure
  • Graduated chambers for easy reference
Specifications
AutoclavableAutoclavable
MaterialPolysulfone
No. per Pack1
Volume (Metric) Receiver1000 mL
Volume (Metric) Upper500 mL
Unit SizeEA