SoftFit-L™ Filtered Pipette Tips in Hinged Racks
Actual product may vary
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
Pipette samples with the accuracy and precision you expect from your pipette, every time with these pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Catalog NumberVolume (Metric)
2749-HR20 μL
Catalog number 2749-HR
HSN 84799090
Price (SGD)
58.50
Special offer
Ends: 30-Nov-2026
195.00
Save 136.50 (70%)
Each
Volume (Metric):
20 μL
Price (SGD)
58.50
Special offer
Ends: 30-Nov-2026
195.00
Save 136.50 (70%)
Each
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
Specifications
Description20 μL, Filtered, Sterile, Hinged Rack
Volume (Metric)20 μL
FilteredFiltered
SterilitySterile
ColorNatural
Model20L
Length (English)1.95 in.
Length (Metric)4.86cm
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal
Color CodeRed
For Use With (Equipment)Rainin LTS LiteTouch
Product LineART SoftFit L
MaterialPolypropylene
Fit for PurposeSterile & PCR Ready
Unit SizeEach