Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Actual product may vary
Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Assure safe centrifugation and storage of PCR reagents, restriction enzymes, and other diagnostic and biochemical reagents or samples by using these storage vials with caps.
Have Questions?
Catalog NumberVolume (Metric) Working
2642620.5 mL
Catalog number 264262
Price (SGD)
-
Volume (Metric) Working:
0.5 mL
Assure safe centrifugation and storage of PCR reagents, restriction enzymes and other biochemical reagents and samples by using Thermo Scientific™ Nunc™ Storage Vials with Caps. Available in 0.5mL and 2.0mL sizes with frosted writing area, the vials have excellent chemical resistance with leakproof closure.
Features:
  • Made from high-purity polypropylene copolymer with excellent chemical resistance
  • Leakproof high-density polyethylene screw closure
  • Centrifuged to a maximum of 13,000 x g
  • Temperature range: -40°C to 95°C
  • RNase/DNase-free
Ordering Information:
Racks 264263 and 270972 are compatible with these vials. Color coders available: 254416, 264335, 264408, 264440, 264459, 264467, 264491.
Specifications
MaterialPPCO
SterilitySterile
Volume (Metric)0.9 mL
No. per Case500
No. per Pack500
Volume (Metric) Working0.5 mL
Product LineNunc
Volume (Metric) Total0.9 mL
Closure MaterialPolyethylene
Closure TypeScrew Cap
Quantity500 Cs.
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeStorage Vial with Cap
For Use With (Equipment)264263, 270972 Rack
Unit SizeCS