Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Actual product may vary
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolumeQuantityUnit Size
254437
also known as NNC254437
10 μLCarton of 600PK
25158610 μLCarton of 1000PK
FIS13-075-4A10 μLCarton of 1000PK
253287
also known as NNC253287
1 μLCarton of 1000PK
FIS13-075-4B1 μLCarton of 1000PK
FKS103447411 μLCarton of 1000PK
NNC2532871 μLCarton of 1000Each
254410
also known as NNC254410
1 μLCarton of 600PK
FIS13-079C1 μLCarton of 600PK
NNC2544101 μLCarton of 600Pack
NNC25443710 μLCarton of 600Case
Catalog number 254437
also known as NNC254437
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume:
10 μL
Quantity:
Carton of 600
Unit Size:
PK
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1 μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
Specifications
DisposableSingle-use
MaterialPolystyrene
SterilitySterile
Volume10 μL
ColorBlue
Product TypeInoculating Loop (Blue)
QuantityCarton of 600
Unit SizePK