Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Actual product may vary
Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Have Questions?
Catalog number 253988
Price (SGD)
727.00
CS
Add to cart
Price (SGD)
727.00
CS
Add to cart
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
Specifications
DisposableSingle-use
MaterialPolystyrene
SterilitySterile
ColorYellow
Product TypeInoculating Needle
QuantityCarton of 1000
Unit SizeCS