Nunc™ 1536-Well Microplates Plates
Nunc™ 1536-Well Microplates Plates
Actual product may vary
Nunc™ 1536-Well Microplates Plates
Screen four times as many samples in a single plate with Thermo Scientifi™ Nunc™ 1536-Well Microplates, which are ideal for high-throughput screening.
Have Questions?
Catalog NumberColor
253614
also known as NNU253614
Clear
Catalog number 253614
also known as NNU253614
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Clear
Maximize high-throughput screening efficiency by using Thermo Scientific™ Nunc™ 1536-Well Microplates, built according to ANSI standards and with the highest quality.
  • Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum signal reflection and minimal crosstalk between wells
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWithout Lid
Array32 x 48
PackagingInner packs of 30
Product LineNunc
ColorClear
Product TypeMicroplate
Skirt StyleLow Flange Skirted
MaterialPolystyrene
Volume (Metric) Well13.4 μL
No. of Wells1536
Volume (Metric) Working1 to 10 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
No. per Case90
No. per Pack30
Quantity90/Cs.
Unit SizeCase