Strep-Tactin™XT Superflow™ resin
Strep-Tactin™XT Superflow™ resin
Actual product may vary
Strep-Tactin™XT Superflow™ resin
Strep-Tactin™XT Superflow™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Have Questions?
Catalog NumberCapacity (Metric)
2401-05001000 mL
Catalog number 2401-0500
Price (SGD)
42.00
PK
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
42.00
PK
Add to cart
  • Strep-Tactin™XT Superflow™ is a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
  • In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
  • The suspension is suitable for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary.
For Research Use Only.
Specifications
Capacity (Metric)1000 mL
TypeResin
Unit SizePK