Utility Trays for Shaker Platforms
Utility Trays for Shaker Platforms
Actual product may vary
Utility Trays for Shaker Platforms
Use these utility trays with 0.5 in. rubber mats with Thermo Scientific™ shakers.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)Dimensions (L x W)
236027Utility tray for 18 x 18 in. platformMaxQ HP 416, 420, 430, 435, 436, 480, 48145.7 x 45.7 cm, 45.7 x 45.7 cm
Catalog number 236027
Price (SGD)
561.00
EA
Add to cart
Description:
Utility tray for 18 x 18 in. platform
For Use With (Equipment):
MaxQ HP 416, 420, 430, 435, 436, 480, 481
Dimensions (L x W):
45.7 x 45.7 cm, 45.7 x 45.7 cm
Price (SGD)
561.00
EA
Add to cart
Use these Thermo Scientific™ Utility trays for Shaker Platforms with 0.5 in. rubber mats.
Thermo Scientific™ utility trays for Shaker platforms have a 0.5 rubber mat to suit individual needs in the laboratory.
Specifications
Dimensions (L x W)45.7 x 45.7 cm, 45.7 x 45.7 cm
Clamps45.7 cm
DescriptionUtility tray for 18 x 18 in. platform
For Use With (Equipment)MaxQ HP 416, 420, 430, 435, 436, 480, 481
Unit SizeEA