7960 Series Thermal Transfer Printer Ribbon
7960 Series Thermal Transfer Printer Ribbon
Actual product may vary
7960 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with excellent smearing resistance and good solvent resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)Diameter (Metric)
236011300 m40 mm25.4 mm
236010300 m170 mm25.4 mm
Catalog number 236011
Price (SGD)
74.00
EA
Add to cart
Length (Metric):
300 m
Width (Metric):
40 mm
Diameter (Metric):
25.4 mm
Price (SGD)
74.00
EA
Add to cart
When printed on the appropriate Brady™ label, this ribbon gives good solvent resistance.
  • Lower for alcohol
  • High print quality
  • Excellent smear resistance
Specifications
ColorBlack
For Use With (Equipment)BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer
MaterialResin
Length (English)984 ft.
Width (English)1.6 in.
Length (Metric)300 m
Width (Metric)40 mm
Diameter (English)1 in.
Diameter (Metric)25.4 mm
FormatOpen-core Ribbon
Certifications/Compliance2011/65/EU and 2015/863
Alternate Part NumberR7960-40X300/O
Unit SizeEA