Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
Actual product may vary
Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
Thermo Scientific
Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
For Use With: Ideal for use when visual inspection of contents is necessary; Autoclavable; Closure Material: Polypropylene; Color: Clear
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2322-00209 L
FIS02-963-10A9 L
FKS106335219 L
2322-005020 L
FKS1000808020 L
Catalog number 2322-0020
HSN 39233090
Price (SGD)
228.00
Special offer
Ends: 30-Jun-2026
350.00
Save 122.00 (35%)
EA
Capacity (Metric):
9 L
Price (SGD)
228.00
Special offer
Ends: 30-Jun-2026
350.00
Save 122.00 (35%)
EA
Easily fill and clean the Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Spigot, which features a wide-mouth opening, a sturdy stainless-steel handle and space-saving rectangular design. Clear construction allows easy viewing of carboy contents. Super tough; impact resistant. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media. Nalgene polycarbonate carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features
  • Molded in graduations in 1 gallon and 5L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Molded-in spigot boss provides strength and leak resistance
  • Clear wall makes this carboy ideal when visual inspection of contents is necessary
  • Follow autoclave instructions provided with the product for proper function; tubing adapter is not autoclavable
  • 20L size has molded-in hand grips in the sidewall for easier handling
  • Autoclavable/Transparent/Graduated
Note:
  • Completely disengage closure threads and disassemble spigot before autoclaving.
  • Follow product insert instructions for autoclaving to ensure proper function.
  • Note: Tubing adaptor is not autoclavable.
  • Repeated autoclaving will shorten the life of polycarbonate carboys.
  • Use pH neutral detergent and rinse completely prior to autoclaving
Includes:
Polypropylene spigot and screw closure
Please recycle the corrugate cartons and LDPE bags in which your Nalgene carboy products are packaged.
Specifications
AutoclavableAutoclavable
Capacity (English)2.4 gal.
Capacity (Metric)9 L
Closure MaterialPolypropylene
Closure Size100 mm
ColorTransparent
DescriptionAutoclavable Clear Rectangular Carboy w/Spigot
Diameter (Metric) Inner Neck86 mm
For Use With (Application)Ideal for use when visual inspection of contents is necessary
Height (Metric) With Closure361 mm
IncludesMolded-in spigot boss w/tubing adapter
Length (Metric)220 mm
MaterialPolycarbonate
No. per Case6/Cs.
No. per Pack1 Ea.
Product TypeCarboy with Spigot
Width (Metric)153 mm
Unit SizeEA
Frequently asked questions (FAQs)
Do you offer replacement carboy spigots?
The standard Nalgene polyproylene quick action spigot (blue handle and body, white threaded ring for connection to carboy) Cat. No. is 6422-0010 or 6422-0010PK. If the original carboy purchased was a solvent-resistant fluorinated carboy, the replacement (mostly white, with a blue cap on the handle) is the Nalgene Quick-Action Tefzel ETFE Spigot, Cat. No. 6432-0010.