ART™ Barrier Low-Retention Pipette Tips
Actual product may vary
ART™ Barrier Low-Retention Pipette Tips
Eliminate sample retention with these specially designed pipette tips.
Have Questions?
Catalog NumberVolume (Metric)ModelTip StyleFit for Purpose
2279-051000 μLART 1000Ergonomic, Low RetentionSterile and PCR Ready
Catalog number 2279-05
HSN 84799090
Price (SGD)
65.40
Special offer
Ends: 30-Nov-2026
218.00
Save 152.60 (70%)
Pack
Volume (Metric):
1000 μL
Model:
ART 1000
Tip Style:
Ergonomic, Low Retention
Fit for Purpose:
Sterile and PCR Ready
Price (SGD)
65.40
Special offer
Ends: 30-Nov-2026
218.00
Save 152.60 (70%)
Pack
Get Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips, which are specifically designed for eliminating sample retention within the pipette tip.
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Tips seal when exposed to potential contaminants
  • Certified RNase, DNase, ATP, and Endotoxin Free
Specifications
DescriptionART 1000, Filtered, Sterile, Rack
Volume (Metric)1000 μL
FilteredFiltered
SterilitySterile
ColorClear
AutoclavableNot Autoclavable
ModelART 1000
Length (English)3.13 in.
Length (Metric)7.9cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatRack
Tip StyleErgonomic, Low Retention
PackagingRack, 8 x 100
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePack