Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Actual product may vary
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Measure more than one volume accurately by using these graduated polyethylene transfer pipettes. Built-in pipette bulb eliminates need for plugging, packaging, and autoclaving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityGraduationsPackaging Type
2257.7 mLNon-sterile1, 2, 3 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FIS13-711-243.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FKS153678133.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FKS123844687.5 mLNon-sterile0.5, 1.0, 1.5, 2.0, 2.5, 3.0 mL
212-20S3.9 mLSterile0.25, 0.50, 0.75, 1 mL20/Bag, 500/Pk., 4000/Cs.
FKS153078235.8 mLSterile0.25, 0.50, 0.75, 1 mLPack of 500
225-1S7.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS22-030-3207.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS13-711-507.7 mLSterile1, 2, 3 mL20/Bag, 500/Pk., 4000/Cs.
FIS13-711-431.5 mLNon-sterile0.1 , 0.2, 0.3 mLBulk 500/Pk 8Pk/Cs 4000/Cs
FKS102917901.5 mLSterile0.1, 0.2, 0.3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FKS1233447813.7 mLNon-sterile1, 2, 3, 4, 5 mLBulk 250/Pk 10Pk/Cs 2500/Cs
Catalog number 225
Price (SGD)
44.00
PAC
Add to cart
Volume (Metric):
7.7 mL
Sterility:
Non-sterile
Graduations:
1, 2, 3 mL
Packaging Type:
Bulk 500/Pk 10Pk/Cs 5000/Cs
Price (SGD)
44.00
PAC
Add to cart
Accurately measure more than one volume by using Thermo Scientific™ Samco™ Graduated Transfer Pipettes. Single-unit design features a built-in pipette bulb that eliminates need for plugging, packaging, and autoclaving while preventing cross-contamination.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications
Bulb Draw (Metric)3.2 mL
DescriptionTransfer Pipette, Large Bulb, Graduated to 1mL, 2mL, 3mL, Transfer Pipette, Large Bulb, Graduated to 1mL, 2mL, 3mL
Diameter (Metric) Outer7.9 mm
MaterialLDPE, LDPE
SterilityNon-sterile
Drops/mL21
GraduatedYes
Graduations1, 2, 3 mL
ColorPink
Product TypeChemically Resistant Natural Rubber Gloves
Size7, 7.5
Packaging TypeBulk 500/Pk 10Pk/Cs 5000/Cs
Volume (Metric)7.7 mL
Length (Metric)15.5 cm
Thickness (Metric)0.52 mm
TypeGraduated Transfer Pipet
DisposableSingle-use
For Use With (Application)Typically used for cleaning and janitorial, manufacturing and assembly, general handling and processing, kitchen work.
Liner MaterialCotton Flocking
LiningLined
Length (Metric)300 mm
Unit SizePAC