Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Actual product may vary
Samco
Samco™ Graduated Transfer Pipettes
Measure more than one volume accurately by using these graduated polyethylene transfer pipettes. Built-in pipette bulb eliminates need for plugging, packaging, and autoclaving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityGraduationsPackaging Type
225-1S7.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS13-711-243.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FKS153678133.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FKS123844687.5 mLNon-sterile0.5, 1.0, 1.5, 2.0, 2.5, 3.0 mL
212-20S3.9 mLSterile0.25, 0.50, 0.75, 1 mL20/Bag, 500/Pk., 4000/Cs.
FKS153078235.8 mLSterile0.25, 0.50, 0.75, 1 mLPack of 500
2257.7 mLNon-sterile1, 2, 3 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FIS22-030-3207.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS13-711-507.7 mLSterile1, 2, 3 mL20/Bag, 500/Pk., 4000/Cs.
FIS13-711-431.5 mLNon-sterile0.1 , 0.2, 0.3 mLBulk 500/Pk 8Pk/Cs 4000/Cs
FKS102917901.5 mLSterile0.1, 0.2, 0.3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FKS1233447813.7 mLNon-sterile1, 2, 3, 4, 5 mLBulk 250/Pk 10Pk/Cs 2500/Cs
222
also known as CA22B2A
5.8 mLSterile, Sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
Catalog number 225-1S
Price (SGD)
68.13
Special offer
Ends: 15-Dec-2025
105.00
Save 36.87 (35%)
PAC
Add to cart
Volume (Metric):
7.7 mL
Sterility:
Sterile
Graduations:
1, 2, 3 mL
Packaging Type:
Individually Wrapped 500/Pk 8Pk/Cs 4000/Cs
Price (SGD)
68.13
Special offer
Ends: 15-Dec-2025
105.00
Save 36.87 (35%)
PAC
Add to cart
Accurately measure more than one volume by using Thermo Scientific™ Samco™ Graduated Transfer Pipettes. Single-unit design features a built-in pipette bulb that eliminates need for plugging, packaging, and autoclaving while preventing cross-contamination.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications
Bulb Draw (Metric)3.2 mL
DescriptionTransfer Pipette, Large Bulb, Graduated to 1mL, 2mL, 3mL
Diameter (Metric) Outer7.9 mm
MaterialLDPE
SterilitySterile
Drops/mL21
GraduatedYes
Graduations1, 2, 3 mL
Packaging TypeIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
Volume (Metric)7.7 mL
Length (Metric)15.5 cm
TypeGraduated Transfer Pipet
DisposableSingle-use
Unit SizePAC