Life Cycle Figurines
Life Cycle Figurines
Actual product may vary
Life Cycle Figurines
Life Cycle Figurines
Life Cycle Figurines
Life Cycle Figurines
Life Cycle Figurines
Life Cycle Figurines
Life Cycle Figurines
Life Cycle Figurines
Life Cycle Figurines
Life Cycle Figurines
STEM tool to learn how different bugs and frogs grow.
Have Questions?
Catalog number 2205
also known as HAB2205
Price (SGD)
31.00
PK
Add to cart
Price (SGD)
31.00
PK
Add to cart
  • High quality figurines are organized in a handy carrying case
  • Life Cycle sorting cards provide additional learning activities
  • Perfect for counting, sequencing, and play
  • Durable and over sided for little hands
  • STEM.org Authenticated Learning Tool can help cultivate an interest in STEM science, technology, engineering, and mathematics
  • True-to-life figurines help kids understand the life cycle of bugs and frogs
Specifications
For Use With (Application)Counting, Sequencing, and Playing
Includes6 life cycle stages- Butterfly, Ladybug, Honey Bee, Praying Mantis, Ant and Frog
Unit SizePK
Frequently asked questions (FAQs)
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?
Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.
For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?
The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.