ART™ Barrier Extended Length Reload System
Actual product may vary
ART™ Barrier Extended Length Reload System
Perform applications accurately and consistently without compromising protection with this barrier reload system.
Have Questions?
Catalog NumberTip StyleFit for Purpose
2179-05-RTLow Retention, Extended LengthSterile and PCR Ready
Catalog number 2179-05-RT
HSN 84799090
Price (SGD)
128.00
Special offer
Ends: 30-Jun-2026
160.00
Save 32.00 (20%)
PK
Tip Style:
Low Retention, Extended Length
Fit for Purpose:
Sterile and PCR Ready
Price (SGD)
128.00
Special offer
Ends: 30-Jun-2026
160.00
Save 32.00 (20%)
PK
Perform applications accurately and consistently without compromising protection with the Thermo ScientificTM ARTTM Barrier Reload System, which provides an environmentally-responsible solution without compromising sterility. With offset wells that allow the tips to stack, this advanced system reduces waste and saves bench top and storage space. The ART portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in your laboratory.
  • ART barrier tips provide 100% security against aerosols and liquid contamination
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert
Specifications
DescriptionART Barrier, Filtered, LR, Sterile, Rack
Volume (Metric)1000 μL
FilteredFiltered
SterilitySterile
ColorNatural
ModelART Barrier
Length (English)3.5 in.
Length (Metric)8.89 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatRack
Tip StyleLow Retention, Extended Length
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePK