Eco Safety Sign, CAUTION Watch For Forklift Traffic
Eco Safety Sign, CAUTION Watch For Forklift Traffic
Actual product may vary
Eco Safety Sign, CAUTION Watch For Forklift Traffic
ZING Enterprises Eco Safety Sign is available in two sizes and three eco-friendly materials.
Have Questions?
Catalog NumberMaterialMounting TypeDimensions (L x W)
2155PlasticCorner Holes10 x 14 in.
Catalog number 2155
Price (SGD)
257.00
PK
Add to cart
Material:
Plastic
Mounting Type:
Corner Holes
Dimensions (L x W):
10 x 14 in.
Price (SGD)
257.00
PK
Add to cart
Plastic signs
  • Made from a minimum of 70% post-consumer recycled materials
  • Printed with UV-resistant inks.
  • Pre-drilled for easy mounting
  • Include adhesive mounting tabs.
  • Impact-resistant plastic is suitable for indoor or outdoor use.
Aluminum signs
  • Made from 94% recycled materials.
  • Printed with UV-resistant inks, and are suitable for indoor and outdoor use.
  • Pre-drilled for easy mounting.
Adhesive signs
  • Made from a minimum of 70% post-industrial polystyrene
  • Resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. Sign meets OSHA and NFPA Life Safety Code standards.
Specifications
MaterialPlastic
Quantity1 Ea.
Mounting TypeCorner Holes
For Use With (Application)Communication
Height (English)10 in.
Width (English)14 in.
LanguagesEnglish
Dimensions (L x W)10 x 14 in.
Displayed TextCAUTION Watch For Forklift Traffic
ColorBlack on Yellow
Unit SizePK