Specifications
DescriptionART 20P, Filtered, Sterile, Hinged Rack
Volume (Metric)20 μL
FilteredFiltered
SterilitySterile
ModelART 20P
Length (English)2 in.
Length (Metric)5.07cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleLow Retention
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePack