Thermo-Flask™ Benchtop Liquid Nitrogen Containers
Actual product may vary
Thermo-Flask™ Benchtop Liquid Nitrogen Containers
Maximize coolant retention with these benchtop liquid nitrogen containers, which contain inner vessels made of borosilicate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Includes
21232 LCarrying Handle, Vented Secure Lid
21221 LCarrying Handle, Vented Secure Lid
FIS11-670-4B2 LCarrying Handle, Vented Secure Lid
FKS118680032 LCarrying Handle, Vented Secure Lid
FIS13-262-7801 LWithout Lid and Handle
FIS13-762-3721.9 LWithout Lid and Handle
Catalog number 2123
HSN 96170090
Price (SGD)
609.00
Special offer
Ends: 30-Jun-2026
676.00
Save 67.00 (10%)
Each
Capacity (Metric):
2 L
Includes:
Carrying Handle, Vented Secure Lid
Price (SGD)
609.00
Special offer
Ends: 30-Jun-2026
676.00
Save 67.00 (10%)
Each
Maximize coolant retention with light-weight Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers, which contain inner vessels of borosilicate and are perfect for everyday cryogenic storage use.
Stainless Steel:
  • Durable, corrosion-resistant stainless-steel exterior
  • Carrying handle and vented secure lid (Part numbers 2122, 2123, 2124)
Shallow Wide Mouth:
  • Ideal for use as a cold trap
  • Convenient dish-shaped design perfect for benchtop use with crushed ice and dry ice/alcohol
  • Stainless-steel jacket and cushioned base for years of trouble-free use
  • Without lid and handle (Part number 2129)
  • Warranty varies by country, please contact us for details
Specifications
TypeBenchtop LN2 Container
Capacity (Metric)2 L
Exterior FinishStainless Steel
StyleBenchtop
IncludesCarrying Handle, Vented Secure Lid
Height (Metric) Exterior265 mm
Diameter (Metric)142 mm
Diameter (Metric) Inside Neck106 mm
Shipping Weight (English)8 lb.
Shipping Weight (Metric)3.6 kg
Unit SizeEach