Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Size |
---|---|---|
2117-1000 | 1000 mL | 120 mm |
FIS02-925-2B | 60 mL | 53 mm |
FIS11-823-30 | 125 mL | 70 mm |
2117-0250 | 250 mL | 70 mm |
FIS11-823-31 | 250 mL | 70 mm |
FIS11-823-32 | 500 mL | 120 mm |
FIS11-823-33 | 1000 mL | 120 mm |
FKS10067820 | 1000 mL | 120 mm |
Protect against breakage and contamination with Thermo Scientific™ Nalgene™; Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure. These make excellent museum jars offering good clarity with excellent chemical resistance. They are lighter in weight than glass and lose only 2% of formaldehyde per year. Not liquid leakproof.
Note: Completely disengage threads or remove cap before autoclaving.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene products are packaged.
.