Nalgene™ Straight-Sided Wide-Mouth Polycarbonate Jars with Closure
Nalgene™ Straight-Sided Wide-Mouth Polycarbonate Jars with Closure
Actual product may vary
Nalgene™ Straight-Sided Wide-Mouth Polycarbonate Jars with Closure
Collect and store pastes, solids, specimens and lab supplies in these clear, straight-sided wide-mouth PC jars, which are high impact-resistant and extremely versatile.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2116-1000
also known as 2116-1000P
1000 mL120 mm
2116-0015
also known as 2116-0015JP
15 mL38 mm
2116-0060
also known as 2116-0060JP
60 mL53 mm
2116-0125125 mL70 mm
2116-0250250 mL70 mm
2116-0500
also known as 2116-0500P
500 mL120 mm
FIS02-925-1H500 mL120 mm
FIS02-925-1J1000 mL120 mm
Catalog number 2116-1000
also known as 2116-1000P
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Closure Size:
120 mm
Collect and store pastes, solids, specimens and lab supplies with window-clear Thermo Scientific™ Nalgene™ Straight-side Wide-Mouth Jars. A wide temperature range (−135°C to +135°C) makes these jars an excellent choice for cold room and refrigerated use all the way to the autoclave. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. These high impact-resistant jars are extremely versatile. Not liquid leakproof.
  • Excellent for cold room and refrigerated use — easy to see contents
  • Wide-mouth and straight sides provide full-access and complete sample retrieval ease
  • 15, 30 and 60mL jars have natural polypropylene closures
  • 125mL to 1L jars have white polypropylene closures with recessed tops for stacking
  • 125 and 250mL sizes fit Osterizer™ and other blenders that have Mason Jar threads; recommended for use with Fluid Transfer Closure DS2153-0700
  • Not liquid leakproof; not recommended for liquid transport
  • Compatible with most weak acids, aliphatic alcohols and aliphatic hydrocarbons.  Not for use with solvents, bases, strong acids, and other harsh laboratory chemicals.  Consult the Nalgene Chemical Resistance guide for more detailed chemical compatibility ratings. 
  • Autoclavable/Transparent
Note: Completely disengage threads or remove cap before autoclaving.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.
Specifications
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorClear
Diameter (English)4.4 in.
Diameter (English) Inner Neck4.4 in.
Diameter (Metric) Inner Neck112 mm
IncludesClosure
MaterialPolycarbonate
No. per Case16
No. per Pack4
Weight (Metric)0.18 kg
ClarityClear
MouthWide
ShapeRound
Height (Metric)151 mm
Product TypeStraight-sided Jar
Height (English)6 in.
Closure MaterialPolypropylene
Closure Size120 mm
Diameter (Metric)112 mm
Closure TypeScrew Cap
Unit SizePK