Specifications
DescriptionART Barrier, Filtered, Sterile, Rack
Volume (Metric)300 μL
FilteredFiltered
SterilitySterile
ColorNatural
ModelART Barrier
Length (English)2.3 in.
Length (Metric)5.84 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatRack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePK