ART™ Wide Bore Filtered Pipette Tips
Actual product may vary
ART™ Wide Bore Filtered Pipette Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Catalog NumberVolume (Metric)Fit for Purpose
2069G200 μLSterile and PCR Ready
Catalog number 2069G
HSN 84799090
Price (SGD)
200.00
Pack
Volume (Metric):
200 μL
Fit for Purpose:
Sterile and PCR Ready
Price (SGD)
200.00
Pack
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Wide Bore Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
  • Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
DescriptionART 200G, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric)200 μL
FilteredFiltered
SterilitySterile
ModelART 200G
Length (English)1.96 in.
Length (Metric)4.97cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatLift-off Lid Rack
Tip StyleGenomic/Wide Orifice
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePack