ART™ Barrier Pipette Tips in Hinged Racks
Actual product may vary
ART™ Barrier Pipette Tips in Hinged Racks
ART™ Barrier Pipette Tips in Hinged Racks
ART™ Barrier Pipette Tips in Hinged Racks
Perform pipetting accurately and consistently with these barrier pipette tips in hinged racks, which seal when exposed to potential contaminants. ART hinged racks are available in low retention, micropoint, extended length and other specialty styles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelFormatTip Style
2069-HR200 μLART 200Hinged RackUniversal Fit
2139-HR10 μLART 10Hinged RackUniversal Fit
FIS02-682-26410 μLART 10Hinged RackUniversal Fit
2139-05-HR10 μLART 10Hinged RackLow Retention
2149P-HR20 μLART 20PHinged RackUniversal Fit
2149P-05-HR20 μLART 20PHinged RackLow Retention
FKS1027694420 μLART 20PHinged RackLow Retention
2065-HR100 μLART 100Hinged RackUniversal Fit
2065E-HR100 μLART 100EHinged RackUniversal Fit
2065E-05-HR100 μLART 100EHinged RackLow Retention
FIS02-682-255200 μLART 200Hinged RackUniversal Fit
2069-05-HR200 μLART 200Hinged RackLow Retention
FKS10768735200 μLART 200Hinged RackLow Retention
2070-HR300 μLART BarrierRackUniversal Fit
FIS13-811-140300 μLART BarrierRackUniversal Fit
2070-05-HR300 μLART BarrierRackLow Retention
2179-HR1000 μLART BarrierHinged RackUniversal Fit
FIS02-682-2681000 μLART BarrierHinged RackUniversal Fit
Catalog number 2069-HR
HSN 84799090
Price (SGD)
36.90
Special offer
Ends: 30-Nov-2026
123.00
Save 86.10 (70%)
Pack
Volume (Metric):
200 μL
Model:
ART 200
Format:
Hinged Rack
Tip Style:
Universal Fit
Price (SGD)
36.90
Special offer
Ends: 30-Nov-2026
123.00
Save 86.10 (70%)
Pack
Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in low retention, micropoint, extended length, and other specialty styles.
Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
DescriptionART 200, Filtered, Sterile, Hinged Rack
Volume (Metric)200 μL
FilteredFiltered
SterilitySterile
ModelART 200
Length (English)2.32 in.
Length (Metric)5.88cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePack