Check Order Status
Six-Piece Jewelers Screwdriver Set
Actual product may vary
Six-Piece Jewelers Screwdriver Set
Three star six-piece jewelers' precision miniature screwdriver set.
Have Questions?
Catalog Number
Head Style
203
Hexagonal
Catalog number
203
Price (SGD)
-
Head Style:
Hexagonal
The 203 three star jewelers' precision miniature screwdriver set is a 6 piece set (0.025 in., 0.030 in., 0.038 in., 0.045 in., 0.055 in., 0.070 in.).
It features nickel-plated handles with revolving, hexagonal heads.
The blades are tempered blue steel.
Specifications
Product Type
Jewelers Screwdriver
Handle Material
Nickel-plated
Head Style
Hexagonal
Material
Tempered Blue Steel
Includes
Blue pouch
Tip Style
Flat