Nalgene™ Narrow-Mouth LDPE Bottles with Closure
Nalgene™ Narrow-Mouth LDPE Bottles with Closure
Actual product may vary
Nalgene™ Narrow-Mouth LDPE Bottles with Closure
Carry out applications such as trace metal analysis with flexible, impact- and chemical-resistant LDPE bottles with closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2003-0004125 mL24 mm
FIS02-924-6B60 mL20 mm
FIS02-924-6C125 mL24 mm
2003-0008250 mL24 mm
FIS02-924-6D250 mL24 mm
2003-0016500 mL28 mm
FIS02-924-6E500 mL28 mm
FIS02-924-6F1 L38-430 mm
FKS103339811 L38-430 mm
Catalog number 2003-0004
Price (SGD)
166.00
CS
Add to cart
Capacity (Metric):
125 mL
Closure Size:
24 mm
Price (SGD)
166.00
CS
Add to cart
Carry out applications such as trace metal analysis with flexible, impact- and chemical-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure, which are extremely low in trace metal content. The bottles are leakproof† when used with Nalgene closures and provide excellent visibility of contents, as well as excellent chemical resistance to most acids, bases and alcohols. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features:
  • Excellent chemical resistance to most acids, bases and alcohols make this a versatile bottle
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Translucence allows easy viewing of liquid levels
  • Linerless caps ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions
  • Naturally low in trace metal content
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Note:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
ColorNatural
DescriptionNarrow-Mouth Bottle
Diameter (Metric) Inner Neck18 mm
MaterialLDPE
No. per Case72
No. per Pack12
SterilityNon-sterile
Closure ColorNatural
Neck Finish24 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)101 mm
Product TypeNarrow Mouth Bottle
Width (English)1.98 in.
Height (English)3.97 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size24 mm
Diameter (Metric)50 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCS
Frequently asked questions (FAQs)
Why are your Nalgene containers and caps always different materials?
In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.