ThermoChill III Recirculating Chillers
ThermoChill III Recirculating Chillers
Actual product may vary
ThermoChill III Recirculating Chillers
Reliable liquid temperature control, with cooling capacity up to 2000w/6830 BTU.
Have Questions?
Catalog NumberCooling CapacityFlow RateCommunicationsPump Type
198222010000Up to 1900 watts9.4 L/min.NonePD2 (Positive Displacement Pump)
Catalog number 198222010000
Price (SGD)
-
Cooling Capacity:
Up to 1900 watts
Flow Rate:
9.4 L/min.
Communications:
None
Pump Type:
PD2 (Positive Displacement Pump)
Reliable liquid temperature control, with cooling capacity up to 2000w/6830 BTU. Thermo Scientific™ ThermoChill Recirculating Chillers are a compact line of refrigerated recirculators ranging in cooling capacities from 600 to 2000 watts. Innovative design features minimized plumbing connections and an integrated full-flow fluid strainer maximize product reliability. With simplicity in design and operation, the ThermoChill platform will provide years of reliable liquid temperature control to meet all of your conventional application requirements.
  • Accessible fill port for easy filling
  • Pressure gauge located on control panel for easy viewing
  • Intuitive digital controller
  • Controller indicator lights illuminate when adjusting settings or when high/low temp alarms activated
  • Removable full-low strainer ensures clean recirculating fluid
  • Locking caster for easy unit maneuvering
  • RS232 or RS485 digital communications optional
Specifications
Description(100 Cell Divider)
Certifications/ComplianceCE marked/UL listed
CommunicationsNone
Cooling CapacityUp to 1900 watts
Flow Rate9.4 L/min.
Pump TypePD2 (Positive Displacement Pump)
Temperature Range-10°C to +30°C
ModelThermoChill III LR
RefrigerantR134A
Temperature (Setpoint)-10°C to +30°C
Temperature Stability±0.1°C
Electrical Requirements230 V/50 Hz