Forensic Toxicology Experiment Kit
Forensic Toxicology Experiment Kit
Actual product may vary
Forensic Toxicology Experiment Kit
Forensic Toxicology Experiment Kit
Forensic Toxicology Experiment Kit
Student's educational kit to analyze crime scene samples for presence of drugs.
Have Questions?
Catalog NumberClass Size
195
also known as BKP195
10 Lab Groups
Catalog number 195
also known as BKP195
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Class Size:
10 Lab Groups
  • Identify drugs, toxins in samples collected from crime scenes, victims, potential suspects
  • Uses Enzyme Linked Immunosorbent Assay (ELISA) to analyze simulated crime scene samples for the presence of drugs
Specifications
Product TypeForensic Toxicology Experiment Kit
Grade9 to Undergraduate
For Use With (Application)Used for educational purposes
IncludesInstructions, simulated crime scene samples, antibodies, buffers, microtiter plates, assorted pipets, microcentrifuge tubes.
Time Range1 Hr.
Class Size10 Lab Groups
Unit SizeEach
Contents & Storage
Refrigeration required