Hot Gas Bypass Temperature Controllers
Hot Gas Bypass Temperature Controllers
Actual product may vary
Hot Gas Bypass Temperature Controllers
Combine these hot gas bypass temperature controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVoltage
195701
also known as GT195701MTK
Field-Installed220 V
195023
also known as GT195023
Factory-Installed220 V
Catalog number 195701
also known as GT195701MTK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Field-Installed
Voltage:
220 V
Combine Thermo Scientific™ Hot Gas Bypass Temperature Controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
Features:
  • 120V or 220V
  • Factory or field installed
Ordering Information:
Field installed option must be installed by a qualified professional.
  • Warranty varies by country, please contact us for details
  • Specifications
    DescriptionField installed (requires qualified professional)
    TypeField-Installed
    Certifications/ComplianceASTM D6319, ASTM D6978-05, ASTM F1671, FDA 21 CFR 177.2600, USP 800
    Certifications/Compliance (Europe)EN455, EU 10/2011 (EN1186) for food handling, EU Certificate 0321.10013-02.E00-00 (Type C, CE 0120)
    Chemotherapeutic RatedYes
    ChlorinatedNo
    Accelerator FreeYes
    ApplicationsHealthcare, Laboratory
    AQL1.5
    ColorBlue
    DisposableSingle-use
    Exterior FinishPartially Textured
    Finger Thickness (English)4.7 mil
    Finger Thickness (Metric)0.12 mm
    Latex FreeYes
    MaterialNitrile
    Palm Thickness (English)2.7 mil
    For Use With (Equipment)CryoPlus 1,2,3,4
    Palm Thickness (Metric)0.07 mm
    PowderedPowder Free
    Product TypeNitrile Examination Glove
    SizeSmall
    SterilityNon-sterile
    Thickness (English)4.7 mil
    Thickness (Metric)0.12 mm
    FDA 510 (k)Yes
    Model701
    SeriesHandPRO™ Scion700™
    Tensile Strength34 MPa
    Length (English)9 in.
    Length (Metric)22.8 cm
    Voltage220 V
    Unit SizeKT