Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Actual product may vary
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Optimize configuration flexibility of 1300 Series A2 Class II Biological Safety Cabinets with these optional stands, UV-lighting, arm rests, foot rests, valves and piping.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
1911313Arm Rest (Set of 2)
1911302Manual Adjustable Stand for 4 ft. Cabinet
FIS15-474-202Manual Adjustable Stand for 4 ft. Cabinet
FIS15-474-203Manual Adjustable Stand for 6 ft. Cabinet
1911306
also known as GT1911306
Vacuum Valve for Universal Piping Option
1911307Water Valve for Universal Piping Option
1911308
also known as FS1911308
Combustible Gas Valve for Standard Applications
1911309
also known as FS1911309
Noncombustible Gas Valve for Standard Applications
FIS15-474-209Noncombustible Gas Valve for Standard Applications
1911310
also known as GT1911310
Vacuum Valve for Standard Applications
FIS15-474-210Vacuum Valve for Standard Applications
1911311Water Valve for Standard Applications
FIS15-474-211Water Valve for Standard Applications
1911312
also known as GT1911312
IV bag holder kit with 12 hooks (for 4 ft., 5 ft., and 6 ft. cabinets)
FIS15-474-212IV bag holder kit with 12 hooks (for 4 ft., 5 ft., and 6 ft. cabinets)
FIS15-474-213Arm Rest (Set of 2)
1911316
also known as FS1911316
Thimble duct exhaust transition for 3 ft., 4 ft., and 5 ft. cabinets
FIS15-474-216Thimble duct exhaust transition for 3 ft., 4 ft., and 5 ft. cabinets
1911317
also known as GT1911317
Thimble Duct Exhaust Transition for 6 ft. Cabinet
FIS15-474-218UV Light Option, Qi
1911319UV light option for 3 ft. and 4 ft. Cabinet
1911321
also known as GT1911321
Universal Exhaust Piping for Right Side of Models 1355 and 1385
1911322Rolling Caster Stand for 4 ft. Cabinet
1911368UV Light for 6 ft. Cabinet
Catalog number 1911313
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Arm Rest (Set of 2)
Utilize these Thermo Scientific™ Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets to increase the flexibility of said cabinets. Accessories include optional stands, UV-lighting, arm rests foot rests, valves and piping.
Stands
  • Manual and electric stands provide comfortable work surface height of 30-37in. (75-95cm) for seated or standing applications
  • Manual stands are adjustable in 1in. increments; electric stands are infinitely adjustable
  • Rolling caster stand provides comfortable fixed worksurface height of 34in. (86cm) for seated applications; designed for limited cabinet mobility for cleaning purposes
  • Stands are included with packages
UV Light
  • Provides safe and easy method for cabinet disinfection
  • Factory-installed; order with cabinet
  • Included with packages
Armrest and Footrest
  • Stainless-steel armrests (set of two) provide ergonomic forearm support and extended reach inside cabinet with less airflow disruption; included with packages
  • Footrest provides ergonomic foot and posture support during seated use
IV Bag Holder Kit
  • Bar and 12 hooks
  • For hanging IV bags near interior ceiling of cabinet
Valves
  • For standard applications or universal piping
  • Standard-application valves have long stem pipe fitting appropriate for side-access media services
  • Short piping stem on universal-piping valves will not penetrate cabinet side wall
  • Combustible gas valve: Rated for combustible gas dispensing inside cabinet chamber
  • Noncombustible gas valve: Rated for noncombustible gas dispensing inside cabinet chamber
  • Vacuum valve: Rated for routing vacuum inside cabinet chamber
  • Water valve: Rated for water dispensing inside cabinet chamber
Thimble Exhaust Duct Transition
  • Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
  • Method recommended by NSF/ANSI™ 49 for externally exhausting Class II, Type A2 biological safety cabinets
Universal Piping for Exhaust Systems
  • Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
  • Factory-installed; must be ordered with cabinet
Universal Piping for Air/Gas Service
  • Allows air and gas service to be piped through top, sides, or bottom of cabinet to minimize side-to-side footprint of cabinet utility piping
  • Two pre-plumbed service ports per side only
  • Factory-installed; must be ordered with cabinet
  • For cabinets with stainless-steel liners, specify using unique part number rather than ordering left and right side separately
Specifications
DescriptionArm Rest (Set of 2)
Certifications/ComplianceNSF/ANSI 49, Class II Type A2
Product TypeArmrest
Height (English)30 in. to 37 in.
For Use With (Equipment)Bio Safety Cabinet
Unit SizeSH