Isotemp™ Support Stand
Product Image
Isotemp™ Support Stand
Material: Steel; For Use With: Napco 8000 Series CO2 Incubators
Have Questions?
Catalog NumberMaterial
190648Steel
Catalog number 190648
Price (SGD)
1,324.00
EA
Add to cart
Material:
Steel
Price (SGD)
1,324.00
EA
Add to cart
  • For Isotemp CO2 Incubators 13-255-25, -26, -27, -28
  • For Napco 8000 Series CO2 Incubators 15-497-001 through -012
Specifications
For Use With (Equipment)Napco 8000 Series CO2 Incubators
LightingLED
MaterialSteel
TypeStand
Height (English)6.5 in.
Height (Metric)16.5 cm
Unit SizeEA