Isotemp™ Support Stand
Isotemp™ Support Stand
Actual product may vary
Isotemp™ Support Stand
Isotemp™ Support Stand
Isotemp™ Support Stand
Isotemp™ Support Stand
Thermo Scientific
Isotemp™ Support Stand
For Use With: Roller Base for WJ Incubator
Have Questions?
Catalog NumberFor Use With (Equipment)
190647Water-Jacketed Incubator
Catalog number 190647
Price (SGD)
1,276.00
EA
Add to cart
For Use With (Equipment):
Water-Jacketed Incubator
Price (SGD)
1,276.00
EA
Add to cart
For Use With: WJ Incubator
  • Roller Base for Water-Jacketed Incubator
Specifications
For Use With (Equipment)Water-Jacketed Incubator
LightingLED
TypeSupport Stand
Height (English)2.75 in.
Height (Metric)7 cm
Unit SizeEA