CD122 Rat anti-Mouse, LE/AF (Low Endotoxin/Azide Free), Clone: 5H4, Southern Biotech™
Product Image
CD122 Rat anti-Mouse, LE/AF (Low Endotoxin/Azide Free), Clone: 5H4, Southern Biotech™
Rat Monoclonal Antibody
Have Questions?
Catalog number 190514
also known as GT190514
View Price:Sign in to see your account pricing. Need an account? Register with us today.
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
AntigenCD122
ApplicationsFlow Cytometry, Immunoprecipitation
ClassificationLow Endotoxin/Azide-free (LE/AF)
Clone5H4
Concentration0.5mg/mL
FormulationSupplied in PBS
Host SpeciesRat
ImmunogenRat myeloma YB2/0 transfected with truncated IL-2Rβ cDNA (YB2/0-mβt-28)
Quantity0.5 mg
Regulatory StatusRUO
Primary or SecondaryPrimary
Target SpeciesMouse
IsotypeIgG2a
Unit SizeKT
Contents & Storage
Store at -20°C