Check Order Status
Access Port
Access Port
For Use With:
Environmental Chambers and Incubators
Have Questions?
Catalog Number
For Use With (Equipment)
190164
also known as MA190164
3962
Catalog number
190164
also known as MA190164
Price (SGD)
861.00
KT
Add to cart
For Use With (Equipment):
3962
Price (SGD)
861.00
KT
Add to cart
Provide a point of entry for probes and monitoring devices in environmental chambers and incubators with Access Ports.
Specifications
Description
Access Port
For Use With (Equipment)
3962
Lighting
LED
Type
Access Port
Installation
Factory, customer specified location
Diameter (Metric) Inside
61 mm
Unit Size
KT