Forma™ Steri-Cult CO2 Incubator Accessories
Forma&trade; Steri-Cult CO<sub>2</sub> Incubator Accessories
Actual product may vary
Forma&trade; Steri-Cult CO<sub>2</sub> Incubator Accessories
Forma&trade; Steri-Cult CO<sub>2</sub> Incubator Accessories
Forma™ Steri-Cult CO2 Incubator Accessories
Increase efficiency, enable portability
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
1900172
also known as GT1900172
Accessory, Shelf RackSteri-Cult CO2 3310 incubator
1900165
also known as GT1900165
Floor StandSteri-Cult CO2s 3307/3308 incubator
1900152
also known as GT1900152
Data OutputSteri-Cult CO2 3307/10 incubator
Catalog number 1900172
also known as GT1900172
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Accessory, Shelf Rack
For Use With (Equipment):
Steri-Cult CO2 3310 incubator
Increase efficiency, enable portability
Specifications
For Use With (Equipment)Steri-Cult CO2 3310 incubator
TypeAccessory, Shelf Rack
Width (English)8.6 in.
Width (Metric)21.8 cm
Height (English)6.6 in.
Height (Metric)16.8 cm
Depth (English)19.8 in.
Depth (Metric)50.29 cm
Unit SizeKT