CO2 Gas Guard
CO<sub>2</sub> Gas Guard
Actual product may vary
CO2 Gas Guard
For Use With: 3307/3308, 3310/3311 Incubators
Have Questions?
Catalog number 1900086
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Minimize the need to frequently change out cylinders and reduce the risk of running out of gas. Thermo Scientific™ Gas Guards for CO2 and LN2 Tanks are factory installed and built in to specified CO2 incubators; specify when ordering the incubator. Fitting unobtrusively inside the incubator casing, these systems interface directly with unit power supply and control systems.
  • Factory-installed systems monitor CO2 or N2 pressure in source tanks
  • Automatically switch from one cylinder to another when supply in first tank is exhausted (pressure falls below 15psi)
  • Audible and visual alarms on incubator control panel alert to tank switch-overs and to when both tanks are low
Specifications
DescriptionGas Guard, CO2
For Use With (Equipment)3307/3308, 3310/3311 incubators
LightingLED
TypeAccessory, CO2 Gas Guard
Unit SizeKT