Tempshield™ Cryo Gloves - Shoulder Length
Actual product may vary
Thermo Scientific
Tempshield™ Cryo Gloves - Shoulder Length
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
189449Large
189448Medium
Catalog number 189449
HSN 40159099
Price (SGD)
835.00
Pair
Size:
Large
Price (SGD)
835.00
Pair
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods. Safety certified to meet EU standards for CE Cat. III: EN511, EN388, EN ISO 21420. Available in wrist, elbow, mid-arm, shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
Specifications
Certifications/ComplianceCE Cat. III: EN 511, EN 388, EN ISO 21420
DescriptionShoulder Length
Product TypeWaterproof Cryogenic Gloves
SizeLarge
For Use With (Application)Cryogenic Preservation
Length (English)28 in.
Length (Metric)71 cm
Unit SizePair