1x1 image pixel for data collection
1500 Series A2 BSC; 6 ft, with SmartCoat or Stainless Steel Interior
1500 Series A2 BSC; 6 ft, with SmartCoat or Stainless Steel Interior
Actual product may vary
Thermo Scientific
1500 Series A2 BSC; 6 ft, with SmartCoat or Stainless Steel Interior
Provides personal, sample and environmental protection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/ComplianceIncludesPlug TypeChamber Material
1589-GNSFAdjustable Stand, UV, ArmrestsUKSmartCoat
1579-GNSFManual Adjustable Height Stand, UV, ArmrestsUKStainless Steel
Catalog number 1589-G
Price (SGD)
16,165.00
EA
Estimated availability date 28-Dec-2025
Add to cart
Certifications/Compliance:
NSF
Includes:
Adjustable Stand, UV, Armrests
Plug Type:
UK
Chamber Material:
SmartCoat
Price (SGD)
16,165.00
EA
Add to cart
1500 Series A2 Biological Safety Cabinets only or cabinet packages. Choice of interior materials. Provides personal, sample and environmental protection. Available in two sizes and fully certified.
  • 1500 Series A2 Biological Safety Cabinet
  • Available certified to NSF & YY
  • Provides personal, sample and environmental protection
  • Available in 2 sizes
  • SmartFlow Plus airflow technology manages protection from contamination while maximizing energy efficiency
  • The touch screen interface is easy to use and clearly displays status ensuring protection that never takes a day off.
Specifications
Certifications/ComplianceNSF
Chamber MaterialSmartCoat
Width (Metric) Interior180 cm
Product TypeType A2 Biosafety Cabinet
Model1500 Series A2
Noise Level<65 dBA
Plug TypeUK
Power Consumption400 W
Receptacles2 Single
Height (English)61.8 in.
Sash Opening (Metric)25 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust878 m3/hr.
For Use With (Application)For Research Only
Width (Metric) Exterior190 cm
IncludesAdjustable Stand, UV, Armrests
Energy Consumption (Reduced Flow Mode)120 W
Height (Metric)156.8 cm
Sash Opening (English)10 in.
Width (English) Interior70.9 in.
Depth (English) Exterior31.5 in.
Depth (Metric) Exterior80 cm
Energy Consumption (Operating Set Point)400 W
Unit SizeEA
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
1579G000424031118Certificate of ComplianceMar 23, 20241579-G
1589G000323102002Certificate of ComplianceNov 04, 20231589-G
1579G000323081709Certificate of ComplianceSep 01, 20231579-G
1579G000323042704Certificate of ComplianceMay 06, 20231579-G
4 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection