1500 Series A2 BSC; 4 ft, with SmartCoat Interior Cabinet Package
Actual product may vary
1500 Series A2 BSC; 4 ft, with SmartCoat Interior Cabinet Package
Choice of interior materials and stand type. factory installed UV light and armrests. Provides personal, sample and environmental protection. Available in 2 sizes fully certified.
Have Questions?
Catalog NumberPlug TypeIncludes
1584-GUKAdjustable Stand, UV, Armrests
Catalog number 1584-G
Price (SGD)
6,300.00
Special offer
Ends: 31-Dec-2026
9,999.00
Save 3,699.00 (37%)
Each
Plug Type:
UK
Includes:
Adjustable Stand, UV, Armrests
Price (SGD)
6,300.00
Special offer
Ends: 31-Dec-2026
9,999.00
Save 3,699.00 (37%)
Each
1500 Series A2 Biological Safety Cabinet package. Choice of interior materials and stand type. factory installed UV light and armrests. Provides personal, sample and environmental protection. Available in 2 sizes fully certified.
  • 1500 Series A2 Biological Safety Cabinet. Available certified to NSF & YY
  • Provides personal, sample and environmental protection
  • Available in 2 sizes
  • SmartFlow Plus airflow technology manages protection from contamination while maximizing energy efficiency
  • The touch screen interface is easy to use and clearly displays status ensuring protection that never takes a day off
Specifications
Certifications/ComplianceNSF
Chamber MaterialSmartCoat
Width (Metric) Interior120 cm
Product TypeType A2 Biosafety Cabinet
Model1500 Series A2
Noise Level<63 dBA
Plug TypeUK
Power Consumption200 W
Receptacles2 Single
Height (English)61.8 in.
Sash Opening (Metric)25 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust582 m3/hr.
For Use With (Application)For Research Only
Width (Metric) Exterior130 cm
IncludesAdjustable Stand, UV, Armrests
Energy Consumption (Reduced Flow Mode)70 W
Height (Metric)156.8 cm
Sash Opening (English)10 in.
Width (English) Interior47.2 in.
Depth (English) Exterior31.5 in.
Depth (Metric) Exterior80 cm
Energy Consumption (Operating Set Point)200 W
Unit SizeEach