1500 Series A2 BSC; 6 ft, with SmartCoat or Stainless Steel Interior
Actual product may vary
Thermo Scientific
1500 Series A2 BSC; 6 ft, with SmartCoat or Stainless Steel Interior
Provides personal, sample and environmental protection.
Have Questions?
Catalog NumberPlug TypeChamber MaterialIncludesCertifications/Compliance
1579-GUKStainless SteelManual Adjustable Height Stand, UV, ArmrestsNSF
Catalog number 1579-G
Price (SGD)
14,747.00
Each
Plug Type:
UK
Chamber Material:
Stainless Steel
Includes:
Manual Adjustable Height Stand, UV, Armrests
Certifications/Compliance:
NSF
Price (SGD)
14,747.00
Each
1500 Series A2 Biological Safety Cabinets only or cabinet packages. Choice of interior materials. Provides personal, sample and environmental protection. Available in two sizes and fully certified.
  • 1500 Series A2 Biological Safety Cabinet
  • Available certified to NSF & YY
  • Provides personal, sample and environmental protection
  • Available in 2 sizes
  • SmartFlow Plus airflow technology manages protection from contamination while maximizing energy efficiency
  • The touch screen interface is easy to use and clearly displays status ensuring protection that never takes a day off.
Specifications
Certifications/ComplianceNSF
Chamber MaterialStainless Steel
Width (Metric) Interior180 cm
Product TypeType A2 Biosafety Cabinet
Model1500 Series A2
Noise Level<65 dBA
Plug TypeUK
Power Consumption400 W
Receptacles2 Single
Height (English)61.8 in.
Sash Opening (Metric)25 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust878 m3/hr.
For Use With (Application)For Research Only
Width (Metric) Exterior190 cm
IncludesManual Adjustable Height Stand, UV, Armrests
Energy Consumption (Reduced Flow Mode)120 W
Height (Metric)156.8 cm
Sash Opening (English)10 in.
Width (English) Interior70.9 in.
Depth (English) Exterior31.5 in.
Depth (Metric) Exterior80 cm
Energy Consumption (Operating Set Point)400 W
Unit SizeEach