1500 Series A2 BSC; 4 ft, with Stainless Steel Interior Cabinets or Cabinet Packages
Actual product may vary
1500 Series A2 BSC; 4 ft, with Stainless Steel Interior Cabinets or Cabinet Packages
1500 Series A2 BSC; 4 ft, with Stainless Steel Interior Cabinets or Cabinet Packages
1500 Series A2 BSC; 4 ft, with Stainless Steel Interior Cabinets or Cabinet Packages
Provides personal, sample and environmental protection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug TypeChamber MaterialIncludesCertifications/Compliance
1574-GUKStainless SteelAdjustable Stand, UV, ArmrestsNSF
1574-AChinaStainless SteelAdjustable Stand, UV, ArmrestsNSF, YY
1574-ACChinaStainless SteelFixed Height Stand, Castors, UV, ArmrestsNSF, YY
Catalog number 1574-G
Price (SGD)
8,533.00
Special offer
Ends: 30-Jun-2026
12,189.00
Save 3,656.00 (30%)
Each
Plug Type:
UK
Chamber Material:
Stainless Steel
Includes:
Adjustable Stand, UV, Armrests
Certifications/Compliance:
NSF
Price (SGD)
8,533.00
Special offer
Ends: 30-Jun-2026
12,189.00
Save 3,656.00 (30%)
Each
1500 Series A2 Biological Safety Cabinets only or cabinet packages. Choice of interior materials. Provides personal, sample and environmental protection. Available in two sizes and fully certified.
  • 1500 Series A2 Biological Safety Cabinet
  • Available certified to NSF & YY
  • Provides personal, sample and environmental protection
  • Available in 2 sizes
  • SmartFlow Plus airflow technology manages protection from contamination while maximizing energy efficiency.
  • The touch screen interface is easy to use and clearly displays status ensuring protection that never takes a day off.
Specifications
Description1500 Series Class II, Type A2 BSC, 4 ft (1.2 m) nominal width with adjustable stand, UV lamp, and armrest
Certifications/ComplianceNSF
Chamber MaterialStainless Steel
Width (Metric) Interior120 cm
Product TypeBiosafety Cabinet
Model1500 Series A2
Noise Level<63 dBA
Plug TypeUK
Power Consumption200 W
Receptacles2 Single
Height (English)61.8 in.
Sash Opening (Metric)25 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust582 m3/hr.
For Use With (Application)For Research Only
Width (Metric) Exterior130 cm
IncludesAdjustable Stand, UV, Armrests
Energy Consumption (Reduced Flow Mode)70 W
Height (Metric)156.8 cm
Sash Opening (English)10 in.
Width (English) Interior47.2 in.
Depth (English) Exterior31.5 in.
Depth (Metric) Exterior80 cm
Energy Consumption (Operating Set Point)200 W
Unit SizeEach