Search Thermo Fisher Scientific
| Catalog Number | Width (English) Cabinet | Chamber Material | Sash Opening (Metric) |
|---|---|---|---|
| 1383 | 5 ft. | Stainless Steel (304) | 25 cm |
Perform daily applications safely and efficiently with simple-to-order Thermo Scientific™ 1300 Series Class II, Type A2 Biological Safety Cabinet Packages, which include: Cabinet; manual, adjustable height stand; factory installed UV light and one set of armrests. These safety cabinets deliver exceptional design and technology advancements such as superior protection with proprietary airflow design, exceptional ergonomics for a safe and comfortable environment, and outstanding energy efficiency for operational cost savings.
Safety Features:
More Choices: