Fixed Volume Labofix Micropipettor
Fixed Volume Labofix Micropipettor
Actual product may vary
Thermo Scientific
Fixed Volume Labofix Micropipettor
Designed to meet the increased demand for fixed volume micropipettes that combine a good quality level with a certainly interesting price. Kartell™ Fixed Volume Labofix Micropipettor is ideal for applications in which a fixed dosage carried out with good precision and accuracy is necessary.
Have Questions?
Catalog NumberVolume (Metric)Compatible Tips
1372
also known as CV137X2
25 μLKartell™ 958/960
Catalog number 1372
also known as CV137X2
Price (SGD)
22,565.00
EA
Add to cart
Volume (Metric):
25 μL
Compatible Tips:
Kartell™ 958/960
Price (SGD)
22,565.00
EA
Add to cart
  • Made of high quality plastic
  • With tip ejector and stainless steel piston for prolonged use over time
  • Handle connection ensures a more comfortable and non-fatiguing grip
  • Tip holder cone can house a wide variety of tips
  • It has a fine shape to make getting inside test tubes or containers having particularly narrow openings or lengthened shapes easier
  • Robust structure of the body ensures high mechanical and chemical resistances
Specifications
Accuracy±0.5%
Certifications/ComplianceEC/IVD, ISO8655
Compatible TipsKartell™ 958/960
AutoclavableAutoclavable
MaterialPlastic
ModelLabofix 25
TypeFixed Volume Micropipettor
Volume (Metric)25 μL
Precision±0.3%
Unit SizeEA