Thermal Mixers and Blocks
Thermal Mixers and Blocks
Actual product may vary
Thermal Mixers and Blocks
Get flexibility with this vertical mixer with variable speed and temperature control for a variety of applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
13687715Block, 20 x 0.5 mL microtubes and 12 x 1.5 mL microtubes
13687711Thermal Mixer
13687712Block, 24 x 1.5 mL microtubes
13687713Block, 24 x 2.0mL microtubes
Catalog number 13687715
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Block, 20 x 0.5 mL microtubes and 12 x 1.5 mL microtubes
Gain variable speed and temperature control with the Thermo Scientific™ Thermal Mixer, which comes with a choice of blocks for microtubes and microplates.
  • Heating and cooling capability to cover a wide range of applications
  • Compact size with quiet operation
  • Convenient packages that include a Thermal Mixer with a Block
Mixer Features
  • Fast heat-up and cooling times and quick to reach specified mixing speed
  • Temperature control range : 15°C below ambient to 100°C
  • Temperature setting range : +4°C to 100°C
  • Shaking speed : 250 to 1400 rpm
  • Temperature stability : ±0.1°C
  • Even mixing across the thermoshaker block
  • Excellent uniformity across the block
  • Digital timer with sound alarm : 1 min to 96 hours
  • Temperature calibration function – compensate for differences in the thermal behavior of different tubes and samples
  • Choice of 5 interchangeable blocks
  • LCD Display indicates the set and actual time of operation, mixing speed, and temperature
  • Heating/Cooling and mixing functions can be run independently of each other
Specifications
Holds20 x 5 mL (Block), 12 x 1.5 mL (Microtubes)
DescriptionBlock, 20 x 0.5 mL microtubes and 12 x 1.5 mL microtubes
No. of Positions20 x 5 mL (Block), 12 x 1.5 mL (Microtubes)
Unit SizeEA