Thermal Mixers and Blocks
Thermal Mixers and Blocks
Actual product may vary
Thermal Mixers and Blocks
Get flexibility with this vertical mixer with variable speed and temperature control for a variety of applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
13687711Thermal Mixer
13687712Block, 24 x 1.5 mL microtubes
13687713Block, 24 x 2.0mL microtubes
13687715Block, 20 x 0.5 mL microtubes and 12 x 1.5 mL microtubes
Catalog number 13687711
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Thermal Mixer
Gain variable speed and temperature control with the Thermo Scientific™ Thermal Mixer, which comes with a choice of blocks for microtubes and microplates.
  • Heating and cooling capability to cover a wide range of applications
  • Compact size with quiet operation
  • Convenient packages that include a Thermal Mixer with a Block
Mixer Features
  • Fast heat-up and cooling times and quick to reach specified mixing speed
  • Temperature control range : 15°C below ambient to 100°C
  • Temperature setting range : +4°C to 100°C
  • Shaking speed : 250 to 1400 rpm
  • Temperature stability : ±0.1°C
  • Even mixing across the thermoshaker block
  • Excellent uniformity across the block
  • Digital timer with sound alarm : 1 min to 96 hours
  • Temperature calibration function – compensate for differences in the thermal behavior of different tubes and samples
  • Choice of 5 interchangeable blocks
  • LCD Display indicates the set and actual time of operation, mixing speed, and temperature
  • Heating/Cooling and mixing functions can be run independently of each other
Specifications
Orbit2 mm
Speed Range250 to 1400 rpm
Voltage120/240 VAC, 12 VDC
Wattage60 W
Amperage4.16 A
Certifications/ComplianceCertificate of Analysis (COA)
Plug TypeUS, EU, UK Plug
Temperature Control15°C below ambient to +100°C
Temperature Setting4°C to 100°C
Temperature Uniformity±0.6°C at 4°C, ±0.1°C at 37°C, ±0.3°C at 100°C
Dimensions (L x W x H)130 x 230 x 205 mm
Height (English)8.07 in.
Height (Metric)205 mm
Length (English)5.11 in.
Length (Metric)130 mm
Width (English)9.05 in.
Width (Metric)230 mm
Shipping Weight (Metric)4 kg
DescriptionThermal Mixer
DisplayLCD
For Use With (Equipment)Thermal Mixer Blocks 13687712, 13687713, 13687714, 13687715, 13687716
Timer Range1 min to 96 hours (1 min increment)
Unit SizeEA